BIOS A and BIOS 16 Centrifuges with GreenCool Technology
Supplier: Thermo Fisher Scientific
75008612 75008614 75008617 75008620 75008621 75008622
77957-138EA 99938 USD
77957-138 77957-140 77957-142 77957-144 77957-146 77957-148
BIOS A and BIOS 16 Centrifuges with GreenCool Technology
Centrifuges Floor-Standing Centrifuges
Thermo Scientific™ BIOS A and BIOS 16 Centrifuges with GreenCool Technology bring enhanced sustainability to bioprocessing facilities and laboratories performing high-throughput applications.
  • GreenCool technology using natural refrigerant (CO2) with a GWP of 1, compliant with F-gas regulations
  • BIOS A models: Up to 10 L capacity, up to 12000×g, BIOS 16 models: Up to 16 L capacity, up to 8500×g
  • Auto-Door™ Function: Open and close the centrifuge door automatically
  • Centri-Touch™ Graphic user interface
  • Thermo Scientific Centri-Cross™ function to convert your existing protocols
  • Auto-Lid™ Function: Instant rotor opening and closing with lid storage
  • Eco-Spin™ Technology using windshielded rotors
  • Fiberlite carbon fiber rotor
GreenCool technology utilizes a natural CO2 refrigerant cooling system that complies with upcoming EU and US EPA F-gas regulations. This refrigerant boasts a Global Warming Potential (GWP) of 1, significantly reducing the environmental impact of centrifuge operation.
Additionally, GreenCool technology offers reduced energy consumption, reducing operational costs, rapid cool-down times, and excellent temperature stability.
Certifications:
UL/CE, Class ISO 6 according to DIN EN ISO 14644-1.