Brady® V3200 Wireless Barcode Scanner, Cradle Barrel Plug Cable, AC Adapter, Power Cord
Supplier: Brady Worldwide
178322
77957-122KT 999.99 USD
77957-122
Brady® V3200 Wireless Barcode Scanner, Cradle Barrel Plug Cable, AC Adapter, Power Cord
Barcode and RFID Systems
Simplify tasks, reduce errors and program to fit only the functions you need with the rugged, reliable V3200 wireless barcode scanner with V1400 cradle and accessories kit.
Delivers consistent, accurate results for any task or skill level, from high-volume warehouse operations to simple data entry. The V1400 supports industrial ethernet, IT (MQTT, OPC UA) and OT (EtherNet/IP, profinet, modbus TCP/IP, BACnet, Siemens S7, Allen-Bradley (CompactLogix™, ControlLogix®, FlexLogix™, MicroLogix™, SLC™, PLC-5E) communication protocols. Cortex decoder technology allows scanner to easily read tiny barcodes as well as shiny and hard-to-read surfaces. Wireless with a 300-foot bluetooth range and 100,000-scan battery capacity for full-shift, on-the-move scanning. Fast wireless charging (0 to 100% in three hours) using the V1400 industrial ethernet bluetooth cradle. Rugged design survives 8-foot drops, harsh environments (IP65 rated) and 250 lbs. of crush force. JavaScript cross-platform compatibility enables a single code base for various devices and seamless scanning. Data parsing sorts and arranges data for your systems to simplify data entry and improve reporting.
Delivery information:
Includes (1) V3200 wireless barcode scanner, (1) V1400 industrial ethernet bluetooth cradle, (1) V1200 Li-ion battery (177234), (1) power and comms connector cable - 5 m (178326), (1) M12 to RJ45 comms cable - 5 m (178331), (1) M12 to barrel plug power cable - 1 m (178327), (1) AC adapter (177241), (1) power cord (177242).