Easily charge, locate and connect your Brady wireless scanner to industrial networks with the V1400 industrial ethernet bluetooth cradle.
- Universal design charges and works with any Brady handheld wireless scanner
Scanner-to-cradle homing helps you easily find misplaced scanners. Supports a wide range of industrial protocols, including EtherNet/IP, PROFINET, Modbus, BACnet and more. Industrial ethernet and IT communication protocols (including MQTT and OPC UA) provide seamless connectivity to PLCs and business systems.
IP 65 rating and rugged design withstand harsh industrial environments and exposure to common chemicals. Easy setup with Rockwell AOP, browser-based configuration and visual guides. LED indicators provide clear status feedback. Tool-free cable disconnect (M12 and RJ45 connections).
Color: blue, gray, yellow
Connectivity options: Bluetooth® 5 Low Energy (class II)
Dimensions: 2.953H×10.04W×4.449" D
Delivery information: Includes (1) V1400 industrial ethernet bluetooth cradle, (1) power and comms connector cable - .5 m (178326), (1) M12 to RJ45 comms cable - 5 m (178331), (1) M12 to barrel plug power cable - 1 m (178327), (1) AC adapter (177241), (1) power cord (177242).