Laboratory Needle Valve Lockout Kit
Supplier: ZING Enterprises
7823
77942-616EA 43 USD
77942-616
Laboratory Needle Valve Lockout Kit
Security Locks
The Laboratory needle valve lockout kit secures four-arm needle valves in lab settings.
  • Locks out four-arm handle needle valves (1 to 2.5") used in labs
  • Durable plastic construction for secure lockout
  • Accommodates up to 3 padlocks (3/8" shackle) for added security
The Laboratory needle valve lockout kit is designed to lock out needle valves with standard four-arm handles typically used in laboratory settings. The device is made in the USA from durable plastic and measures 1"H×3"D. The lockout fits 1 to 2.5" valves and accommodates 3 padlocks with shackles up to ³/₈" diameter.
Ordering information:
Complete lockout kit includes: (1) #7103 handle lockout device, (1) #7308 carrying pouch, (1) #7030 red lockout padlock, (2) #7343 lockout tags, and (2) cable ties.