Laboratory Ball Valve Lockout Kit
Supplier: ZING Enterprises
7822
77942-612EA 49.5 USD
77942-612
Laboratory Ball Valve Lockout Kit
Security Locks
The Laboratory ball valve lockout kit secures lever-handle ball valves in lab settings.
  • Complete lockout solution for laboratory ball valves with lever handles
  • Durable PETG lockout device with a 1" opening
  • Accommodates a single padlock (up to 5/16" shackle)
The Laboratory ball valve lockout kit is designed to lock out ball valves with lever handles typically used in laboratory settings. The device is made from durable red PETG plastic and has a 1" diameter opening that fits over the valve body. It accommodates a single padlock with up to a ⁵/₁₆" diameter shackle.
Ordering information:
Kit includes: (1) #7821 lockout device, (1) #7308 carrying pouch, (1) #7030 red lockout padlock, (2) #7343 lockout tags, and (2) cable ties.