Kidde Pro-5MP Fire Extinguisher, with Wall Hanger, 3-A, 40-B:C
Supplier: Janitorial Supplies
KID21032142
77929-484EA 70 USD
77929-484
Kidde Pro-5MP Fire Extinguisher, with Wall Hanger, 3-A, 40-B:C
First Aid Supplies Fire Extinguishers
Stop common fires involving trash, wood, paper, liquids, gases, and electrical equipment.
  • All-metal construction with metal valve assembly
  • 3× more firefighting power
  • Rechargeable
  • UL rating: 3-A, 40-B:C
  • Weight: 5.5 lbs.
Safeguard any property with the dependable Kidde PRO-5MP rechargeable fire extinguisher, designed for commercial use. This extinguisher is designed to meet code compliance for larger areas and common fires involving wood, paper, and textiles (Class A), flammable liquids and gases (Class B), and electrical equipment (Class C). Manufactured from durable, corrosion-resistant aluminum, the Kidde Pro fire extinguisher is a breeze to handle. It provides a remarkable discharge rate of 13 to 15 seconds, a range of 12 to 18 feet, and an operating pressure of 195psi.
The sleek, painted aluminum handle, aluminum cylinder, and metal valve are impact-resistant, making it dependable when needed most. The easy-to-pull safety pin provides quick access for simple operation designed to combat fire hazards using the PASS (Pull, Aim, Squeeze, Sweep) firefighting method. The multi-directional discharge hose allows action from various angles. Note that the dry chemical agent in this fire extinguisher is non-toxic but may cause skin irritation. Place a fire extinguisher within easy reach where fires are more likely to start. Rechargeable.