Five Star® Composition Book, Medium/College Rule

Supplier: Janitorial Supplies
MEA850065
77929-436PK 9.65 USD
77929-436
Five Star® Composition Book, Medium/College Rule
Notebooks

A Five Star® Composition Book is perfect for permanent notetaking. Use it in class and at home for labs, take-home assignments, and more.



  • Composition book features wireless sewn binding to keep pages permanently bound

  • Durable, plastic cover lasts all year and lies flat when opened
  • Class schedule printed inside
  • Randomly assorted cover color
  • (80) 9.75×7.5" sheets


These composition books are made to be durable, eliminating the worry about binding coming apart or missing pages that have fallen out. Pages are sewn in so they stay in place securely, all year long. The composition book front and back covers are plastic to resist everyday wear and tear. Use with Five Star® Study App to scan, study, and organize notes. Create instant flashcards and sync notes to Google Drive to access them anywhere from any device.


Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR