Energizer® Industrial Lithium CR123 Photo Batteries, 3V
Supplier: Janitorial Supplies
EVEELN12312
77929-402PK 73.25 USD
77929-402
Energizer® Industrial Lithium CR123 Photo Batteries, 3V
Batteries
Power specialty professional devices with Energizer Industrial Lithium 123 batteries.
  • Keep specialty power supply stocked
  • Protect professional equipment with lithium batteries that are 100% leakproof in standard usage
  • Get remarkable performance, even in extreme work site temperatures
Trust your precision electronics to Lithium 123 batteries that are 100% leakproof in standard usage. They operate in worksite temperatures ranging from −40 to 140° F. They hold their power with up to 10 years of shelf life, virtually eliminating device downtime. When work depends on motion sensors, security cameras, two-way radios, and more, trust the power and protection of Energizer Industrial Lithium 123 batteries.