Large Lab Wipe
Supplier: StatLab
SL5790
77908-138PK 12.23 USD
77908-138
Large Lab Wipe
Wipes Disposable Wipes and Towels Wipes for General Applications
Use as an absorbent work surface for coverslipping or for general lab cleanup.
  • Low-linting
  • Heavy-duty, air-layered material