TissueWrap Biopsy Paper
Supplier: StatLab
EBB-0102-10A
77908-038PK 81.54 USD
77908-038
TissueWrap Biopsy Paper
Biopsy Pads
TissueWrap Biopsy Paper is specifically designed for processing biopsy specimens.
  • Tissue paper is cut to a convenient, ready-to-use size
This non-absorbent paper allows fluids to flow through freely while retaining small tissue fragments.