Cotton Tip Applicator Stick
Supplier: StatLab
CTA-806W
77907-984BX 32.06 USD
77907-984
Cotton Tip Applicator Stick
Swabs Swabs for Clinical Use
For application of tissue marking dyes and other routine uses in paraffin and frozen sectioning.
  • Splinter-free applicator stick with cotton tip
  • 6" length
  • Non sterile