Spirochete Control Slides
Supplier: StatLab
CS-SPIRO/25
77907-974PK 364.29 USD
77907-974
Spirochete Control Slides
Slides Microscopy Control Slides
Spirochete control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
  • Includes one stained slide
  • Tissue: known positive
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90° corners. Each package includes specific staining protocol.