Mast Cell IHC Control Slides
Supplier: StatLab
CSM0125P
77907-930PK 453.34 USD
77907-930
Mast Cell IHC Control Slides
Slides Microscopy Control Slides
Mast Cell IHC control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
  • Stained slide is not included
  • Tissue: known positive
  • Special stain or antibody: Mast cell
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90 degree corners. Each pack includes specific staining protocol.