Iron (Liver) (Human) Control Slide
Supplier: StatLab
CS-IRON(L)/25
77907-928PK 207.31 USD
77907-928
Iron (Liver) (Human) Control Slide
Slides Microscopy Control Slides
Iron (Liver) (Human) control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
  • Includes one stained slide
  • Tissue: Human known positive
  • Special stain or antibody: Iron stain
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90° corners. Each package includes specific staining protocol.