Cincinnati Surgical Blade Handle, #4
Supplier: StatLab
CS-H4
77907-922EA 38.29 USD
77907-922
Cincinnati Surgical Blade Handle, #4
Scalpels and Scalpel Assemblies Scalpel Handles
Manufactured from special cold-formed nickel steel.
  • Handles are stainless steel
  • Non sterile
Each handle is designed to incorporate a satin finish, smooth loading, and balance. Cincinnati surgical blade handles are designed for use with Cincinnati surgical blades. It's important to ensure that the right blade is paired with the right handle.