Candida Albicans Control Slide
Supplier: StatLab
CSC0525P
77907-886PK 689.71 USD
77907-886
Candida Albicans Control Slide
Slides Microscopy Control Slides
Candida Albicans control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
  • Stained slide is not included
  • Tissue: known positive
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90° corners. Each package includes specific staining protocol.