Cincinnati Surgical Blades, #70
Supplier: StatLab
CS-B70
77907-882BX 212.34 USD
77907-882
Cincinnati Surgical Blades, #70
Scalpels and Scalpel Assemblies Scalpel Blades
Precision carbon blades manufactured of high-grade surgical carbon steel resulting in exceptional durability and sharpness.
  • Blades are packed 100 per boX
  • Non-sterile Individually wrapped
Cincinnati surgical blades are designed for optimal performance with Cincinnati surgical blade handles. It's important to ensure that the right blade is paired with the right handle. Size 60 and 70 blades (CS-B60, CS-B70) should be paired with a #8 handle (CS-H8). CS-B70 features a blunt tip.