Cincinnati Surgical Blade, #22
Supplier: StatLab
CS-B22
77907-880BX 103.8 USD
77907-880
Cincinnati Surgical Blade, #22
Scalpels and Scalpel Assemblies Scalpel Blades
Precision carbon blades manufactured of high-grade surgical carbon steel resulting in exceptional durability and sharpness.
  • Blades are packaged 100 per box
  • Non sterile
  • Individually wrapped
Cincinnati surgical blades are designed for optimal performance with Cincinnati surgical blade handles. It's important to ensure that the right blade is paired with the right handle. Size 22 blades (CS-B22) should be paired with a #4 handle (CS-H4).