Amyloid (Human) Control Slide
Supplier: StatLab
CS-AMYL/25
77907-874PK 437.14 USD
77907-874
Amyloid (Human) Control Slide
Slides Microscopy Control Slides
Amyloid (Human) control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
  • Includes one stained slide
  • Tissue: known positive special stain
  • Antibody: Congo red amyloid
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using millennia adhesion white glass slides with 90° corners. Each package includes specific staining protocol.