Amyloid Control Slides
Supplier: StatLab
CSA0425P
77907-864PK 598.26 USD
77907-864
Amyloid Control Slides
Slides Microscopy Control Slides
Amyloid Control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements for amyloid.
  • Control slide tissue is cut at 8 microns
  • Intended for use with Congo Red Amyloid Special Stains
  • Not for use with IHC
Includes 25 unstained slides.