Bio-Mesh™ Biopsy Bags
Supplier: StatLab
BBMD1000 BBSM1000
77907-816CS 424.06 USD
77907-816 77907-818
Bio-Mesh™ Biopsy Bags
Bags Sample Bags
Resistent to solvents and acids.
  • Secures small biopsies during processing
  • 1000 bags per case
Synthetic mesh, small pore design, ensures unrestricted flow of processing reagents around specimens.