LDK378 98%

Supplier: Ambeed
Danger

Synonyms: LDK-378, 5-Chloro-N2-(2-isopropoxy-5-methyl-4-(piperidin-4-yl)phenyl)-N4-(2-(isopropylsulfonyl)phenyl)pyrimidine-2,4-diamine, Ceritinib (free base), 5-Chloro-N2-(2-isopropoxy-5-methyl-4-(piperidin-4-yl)phenyl)-N4-(2-(isopropylsulphonyl)phenyl)pyrimidine-2,4-diamine

A126440-10MG A126440-1G A126440-250MG A126440-5G A126440-5MG
77149-438EA 6 USD
77149-438 77549-502 77571-580 77611-086 77901-536
LDK378 98%
LDK378

Caution: for Research Use Only

Formula: C₂₈H₃₆ClN₅O₃S
MW: 558.1 g/mol
MDL Number: MFCD26142648
CAS Number: 1032900-25-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR