General Purpose, Unstirred Water Baths
Supplier: Polyscience
WBE02A11B WBE05A11B WBE10A11B WBE20A11B WBE28A11B
77900-458EA 601.75 USD
77900-458 77900-460 77900-462 77900-464 77900-466
General Purpose, Unstirred Water Baths
Baths Water Baths, Unstirred
Premium digital general-purpose water baths are designed for accurate temperature control in laboratory and industrial applications. Ideal for maintaining consistent temperatures for samples, they are used in research, medical and food processing applications.
  • Five presets for frequently used temperatures
  • Eco-mode for heat-off use
  • Calibration offset allows user to calibrate one temperature point, as needed
  • Safety thermostat and alarms
  • Operates in five languages: Chinese, English, French, German and Spanish
  • Optional use with bath beads, to 40 °C
  • Large buttons are easy to use with gloves
  • Two year parts and labor warranty
These general purpose water baths are designed with the user in mind featuring easy-to-use controls, full-color TFT display, high gabled lids with anti-drip design and durable construction ensuring reliable performance for your heating and incubation needs.
Certifications:
cETLus.
Delivery information:
Supplied with lid and power cord.