Cleanroom Glove Box Dispenser
Supplier: PALBAM 2008 AC LTD TE
GD0208
77898-338EA 249.22 USD
77898-338
Cleanroom Glove Box Dispenser
Glove Dispensers and Glove Box Holders
Palbam’s cleanroom glove box dispenser is a wall-mountable, stainless steel unit designed for cleanrooms. It ensures easy, contamination-free access to gloves, promoting organization and compliance.
  • Stainless steel 304 with electropolished finish
  • Wall mounted
The gownroom is often the first impression for employees and visitors alike. Palbam Class products are guaranteed to ensure that your company will give the best possible impression, as well as serving all ergonomic, cleanliness and functional requirements.