Cleanroom Wipe Down Table
Supplier: PALBAM 2008 AC LTD TE
WDT-0960
77898-322EA 4546.21 USD
77898-322
Cleanroom Wipe Down Table
Furniture Tables and Workbenches
Palbam’s cleanroom wipe down table is a stainless steel, ergonomic solution designed for cleanrooms.
  • Stainless steel 304 with electropolished finish
  • Includes hole in table top for waste disposal
  • Includes removable plastic bin
It provides a durable, contamination-free surface for efficient wipe-down procedures, ensuring compliance and cleanliness in controlled environments. The wipe down table supports the wipe-down procedure perfectly, featuring a hole in the tabletop for easy waste disposal. The cabinet beneath the table and waste disposal hole includes a removable plastic bin for convenient waste management.
The gownroom is often the first impression for employees and visitors alike, that they are entering into a special area. Palbam Class products are guaranteed to ensure that your company will give the best possible impression, as well as serving all the ergonomic, cleanliness and functional requirements.
Ordering information:
Available in several dimensions; contact your Avantor representative for more information.