Eye Glasses Dispenser
Supplier: PALBAM 2008 AC LTD TE
GD-0302
77898-240EA 643.5 USD
77898-240
Eye Glasses Dispenser
PPE Dispensers
Palbam’s wall mounted eyeglasses dispenser is a high-quality, stainless steel unit designed for cleanroom environments. It provides a contamination-free, organized solution for dispensing safety glasses, ensuring easy access and compliance.
  • Stainless steel 304 with electropolished finish
  • Open top and acrylic front panel
  • Can also be used as a small general-purpose dispenser
  • Wall mounted
The gown room is often the first impression for employees and visitors alike. Palbam Class products are guaranteed to ensure that your company will give the best possible impression, as well as serving all ergonomic, cleanliness, and functional requirements.