Bisdemethoxycurcumin ≥98%

Supplier: Ambeed
Warning

Synonyms: 1,7-Bis(4-hydroxyphenyl)-1,6-heptadiene-3,5-dione, 1,7-Bis(4-hydroxyphenyl)hepta-1,6-diene-3,5-dione, Didemethoxycurcumin, Bisdemethoxycucurmin

A606809-5G A606809-25G A606809-1G A606809-100G
77613-880EA 32.96 USD
77613-880 77686-532 77686-531 77893-223
Bisdemethoxycurcumin ≥98%
Bisdemethoxycurcumin

Caution: for Research Use Only

Formula: C₁₉H₁₆O₄
MW: 308.33 g/mol
MDL Number: MFCD03419284
CAS Number: 24939-16-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR