2,7-Diamino-9,9-di-n-octylfluorene 98%

Supplier: Ambeed
Warning

A391620-250MG A391620-1G A391620-100MG
77016-194EA 23 USD
77016-194 77613-182 77890-806
2,7-Diamino-9,9-di-n-octylfluorene 98%
2,7-Diamino-9,9-di-n-octylfluorene

Caution: for Research Use Only

Formula: C₂₉H₄₄N₂
MDL Number: MFCD27923070
CAS Number: 851042-10-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR