AS-703026 98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: (S)-N-(2,3-Dihydroxypropyl)-3-((2-fluoro-4-iodophenyl)amino)isonicotinamide, MSC1936369B, Pimasertib

A199908-10MG A199908-50MG A199908-5MG A199908-1MG A199908-250MG A199908-1G A199908-100MG A199908-5G
77012-608EA 15 USD
77012-608 77012-610 77012-612 77611-924 77611-926 77683-555 77890-206 77890-207
AS-703026 98%, Ambeed.Inc
AS-703026

Caution: For research use only.

Formula: C₁₅H₁₅FIN₃O₃
MDL Number: MFCD16660676
CAS Number: 1236699-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR