VACUUBRAND® RZ 6 Rotary Vane Vacuum Pump
Supplier: Brandtech
20798136
77889-136EA 4500 USD
77889-136
VACUUBRAND® RZ 6 Rotary Vane Vacuum Pump
Pumps Vacuum Rotary Vane Pumps
Compact and powerful, the RZ 6 rotary vane vacuum pump achieves low ultimate vacuum while handling medium to high gas flow rate.
  • Ideal for degassing, lyophilization, fine vacuum applications, and drying chambers
The RZ 6 is a high-performance, two-stage rotary vane pump designed for reliable vacuum generation. Its large oil capacity extends maintenance intervals, while the telescopic design allows for easy servicing. An efficient gas ballast design ensures high water vapor tolerance without compromising vacuum performance. 
Certifications:
NRTL.
Delivery information:
Supplied with pump, filled with oil and completely mounted, includes manual.