TSG Sample Preparation Refrigerators
Supplier: Thermo Fisher Scientific
TSG2305CA TSG3005CA TSG4505CA TSG5005CA
77889-058EA 9811.3 USD
77889-058 77889-060 77889-062 77889-064
TSG Sample Preparation Refrigerators
Refrigerators
TSG Series Sample Prep refrigerators equipped with a duplex outlet and access ports are built to help meet your temperature-controlled sample preparation needs, offering quiet operation, outstanding performance, and sustainable design.
  • Safety switch protected internal electrical duplex outlet
  • Four 2" casters; front casters are lockable
  • Keyed door locks, compatible with popular E-lock adapters
  • ENERGY STAR–certified refrigerator
  • Self-closing doors with 90° stop
  • Double-pane glass doors
  • Bright LED interior lighting and heat-free defrost
The interior convenience outlet offers flexibility and easy setup of benchtop equipment within the chamber. Shake, spin, and agitate samples in a temperature-controlled environment without investing in specialty benchtop equipment. Temperature, door ajar, and power failure alarms. 12-hour controller battery backup and remote alarm contacts and rear access port. Quiet operation at less than 53 dB.
Single and double door sizes ranging from 23 to 49 cu. ft. Two 5 cm (2") side wall access ports with insulated hinged covers for solvent, waste, and other common tubing and hoses.