Methyl 2-acetamido-2-deoxy-α-D-galactopyranoside 95%

Supplier: AFG Bioscience

Synonyms: Methyl a-N-acetyl-D-galactosamine

C54050-1G C54050-2G C54050-500MG C54050-250MG
76620-018EA 300.46 USD
76620-018 76620-020 76620-016 77888-972
Methyl 2-acetamido-2-deoxy-α-D-galactopyranoside 95%
Methyl 2-acetamido-2-deoxy-α-D-galactopyranoside
Formula: C₉H₁₇NO₆
Storage Temperature: Refrigerator
MDL Number: MFCD00077378
CAS Number: 6082-22-0

Order Now

Specification Test Results

Purity Limit 95%
Appearance white powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR