2-O-Methyl-α-D-N-acetylneuraminic acid ≥95%

Supplier: AFG Bioscience

Synonyms: N-Acetyl-2-O-methyl-a-D-neuraminic acid

C53621-100MG C53621-25MG C53621-50MG C53621-10MG
76619-940EA 845.81 USD
76619-940 76619-936 76619-938 77888-927
2-O-Methyl-α-D-N-acetylneuraminic acid ≥95%
2-O-Methyl-α-D-N-acetylneuraminic acid
Formula: C₁₂H₂₁NO₉
Storage Temperature: Freezer
MDL Number: MFCD00084809
CAS Number: 50930-22-8

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White To Off-White Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR