α-Man-PEG3-Alkyne 95%
Supplier: AFG Bioscience
Synonyms:
2-{2-[2-(Prop-2-ynyloxy)ethoxy]ethoxy}ethyl α-D-Mannopyranoside
C52005-50MG
C52005-25MG
C52005-100MG
C52005-10MG
76619-734EA
492.92
USD
76619-734
76619-732
77888-831
77888-832
α-Man-PEG3-Alkyne 95%
α-Man-PEG3-Alkyne
Formula:
C₁₅H₂₆O₉ Storage Temperature: Freezer |
CAS Number:
1442747-33-2 |
Specification Test Results
Purity Limit | 95% |
Appearance | White powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...