β-GlcNAc-PEG3-Alkyne 95%

Supplier: AFG Bioscience

Synonyms: 2-{2-[2-(Prop-2-ynyloxy)ethoxy]ethoxy}ethyl 2-acetamido-2-deoxy-β-D-glucopyranoside, N-((2R,3R,4R,5S,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-2-(2-(2-(2-(prop-2-yn-1-yloxy)ethoxy)ethoxy)ethoxy)tetrahydro-2H-pyran-3-yl)acetamide

C52002-25MG C52002-50MG C52002-100MG C52002-10MG
76619-720EA 295.75 USD
76619-720 76619-722 77888-825 77888-826
β-GlcNAc-PEG3-Alkyne 95%
β-GlcNAc-PEG3-Alkyne
Formula: C₁₇H₂₉NO₉
Storage Temperature: Freezer
CAS Number: 1884184-44-4

Order Now

Specification Test Results

Purity Limit 95%
Appearance White powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR