α-GalNAc-PEG3-alkyne 95%

Supplier: AFG Bioscience

Synonyms: 2-{2-[2-(Prop-2-ynyloxy)ethoxy]ethoxy}ethyl 2-acetamido-2-deoxy-α-D-galactopyranoside

C52001-25MG C52001-50MG C52001-100MG C52001-10MG
76619-716EA 450.69 USD
76619-716 76619-718 77888-823 77888-824
α-GalNAc-PEG3-alkyne 95%
α-GalNAc-PEG3-alkyne

Order Now

Specification Test Results

Purity Limit 95%
Appearance White powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR