Methyl 2,​3,​4-​tri-​O-​acetyl-​β-​D-​glucopyranuronosyl azide ≥95%

Supplier: AFG Bioscience

Synonyms: Methyl 2,3,4-tri-O-acetyl-1-azido-1-deoxy-β-D-glucopyranosyluronate, 1-Azido-2,3,4-tri-O-acetyl-b-D-glucuronide methyl ester, 2,3,4-Tri-O-acetyl-1-azido-1-deoxy-β-D-glucuronic acid methyl ester

C51679-1G C51679-2G C51679-500MG C51679-100MG C51679-250MG
76619-654EA 500.76 USD
76619-654 76619-656 76619-652 77888-797 77888-798
Methyl 2,​3,​4-​tri-​O-​acetyl-​β-​D-​glucopyranuronosyl azide ≥95%
Methyl 2,​3,​4-​tri-​O-​acetyl-​beta-​D-​glucopyranuronosyl azide
Formula: C₁₃H₁₇N₃O₉
Storage Temperature: Refrigerator
MDL Number: MFCD08703845
CAS Number: 67776-38-9

Order Now

Specification Test Results

Purity Limit > 95%
Appearance Pale Brown Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR