5-​Azidopentyl 2-​acetamido-​3,​4,​6-​tri-​O-​acetyl-​2-​deoxy-​β-​D-​galactopyranoside 95%

Supplier: AFG Bioscience

C51615-100MG C51615-250MG C51615-500MG C51615-25MG
76619-636EA 499.98 USD
76619-636 76619-638 76619-640 77888-788
5-​Azidopentyl 2-​acetamido-​3,​4,​6-​tri-​O-​acetyl-​2-​deoxy-​β-​D-​galactopyranoside 95%
5-​Azidopentyl 2-​acetamido-​3,​4,​6-​tri-​O-​acetyl-​2-​deoxy-​β-​D-​galactopyranoside
Formula: C₁₉H₃₀N₄O₉
Storage Temperature: Refrigerator
CAS Number: 1479084-10-0

Order Now

Specification Test Results

Purity Limit 95%
Appearance colorless sticky oil


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR