β-D-GlcNAc-PEG3-Azide, 3,​4,6-​triacetate 95%

Supplier: AFG Bioscience

Synonyms: (2-​(2-​(2-​Azidoethoxy)​ethoxy)​ethyl)​-​2-​acetamido-​2-​deoxy-​3,​4,​6-​tri-​O-​acetyl-​beta-​D-​glucopyranoside

C51603-250MG C51603-500MG C51603-100MG C51603-50MG
76619-628EA 751.14 USD
76619-628 76619-630 77888-778 77888-779
β-D-GlcNAc-PEG3-Azide, 3,​4,6-​triacetate 95%
β-D-GlcNAc-PEG3-Azide, 3,​4,6-​triacetate
Formula: C₂₀H₃₂N₄O₁₁
Storage Temperature: Refrigerator
CAS Number: 86520-53-8

Order Now

Specification Test Results

Purity Limit 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR