6-Azido-6-deoxy-D-galactose peracetate ≥95%

Supplier: AFG Bioscience

Synonyms: 1,2,3,4-Tetra-O-acetyl-6-azido-6-deoxy-D-galactopyranose

C51528-1G C51528-500MG C51528-100MG C51528-250MG
76619-508EA 640.8 USD
76619-508 76619-506 77888-725 77888-726
6-Azido-6-deoxy-D-galactose peracetate ≥95%
6-Azido-6-deoxy-D-galactose peracetate
Formula: C₁₄H₁₉N₃O₉
Storage Temperature: Refrigerator
CAS Number: 629620-22-0

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR