6-Azido-6-deoxy-D-galactose peracetate ≥95%
Supplier: AFG Bioscience
Synonyms:
1,2,3,4-Tetra-O-acetyl-6-azido-6-deoxy-D-galactopyranose
C51528-1G
C51528-500MG
C51528-100MG
C51528-250MG
76619-508EA
640.8
USD
76619-508
76619-506
77888-725
77888-726
6-Azido-6-deoxy-D-galactose peracetate ≥95%
6-Azido-6-deoxy-D-galactose peracetate
Formula:
C₁₄H₁₉N₃O₉ Storage Temperature: Refrigerator |
CAS Number:
629620-22-0 |
Specification Test Results
Purity Limit | > 95% |
Appearance | White Powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...