N-azidoacetylmannosamine 95%

Supplier: AFG Bioscience

Synonyms: ManNAz, 2-[(2-Azidoacetyl)amino]-2-deoxy-D-mannose

C51519-100MG C51519-50MG C51519-10MG C51519-25MG
76619-468EA 993.69 USD
76619-468 76619-466 77888-717 77888-718
N-azidoacetylmannosamine 95%
N-azidoacetylmannosamine
Formula: C₈H₁₄N₄O₆
Storage Temperature: Freezer
CAS Number: 361154-23-6

Order Now

Specification Test Results

Purity Limit 95%
Appearance white powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR