3,4,6-Tri-O-benzyl-2-deoxy-D-glucopyranose ≥95%

Supplier: AFG Bioscience

C50720-1G C50720-5G C50720-250MG
76619-206EA 445.98 USD
76619-206 76619-208 77888-656
3,4,6-Tri-O-benzyl-2-deoxy-D-glucopyranose ≥95%
3,4,6-Tri-O-benzyl-2-deoxy-D-glucopyranose
Formula: C₂₇H₃₀O₅
Storage Temperature: Refrigerator
MDL Number: MFCD06797170
CAS Number: 132732-60-6

Order Now

Specification Test Results

Purity Limit > 95%
Appearance Powder or Crystals


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR