N-Acetylallolactosamine ≥95%

Supplier: AFG Bioscience

Synonyms: β-D-Gal-(1→6)-D-GlcNAc

C50364-100MG C50364-25MG C50364-10MG C50364-5MG
76619-108EA 1352.06 USD
76619-108 76619-106 77888-642 77888-643
N-Acetylallolactosamine ≥95%
N-Acetylallolactosamine
Formula: C₁₄H₂₅NO₁₁
Storage Temperature: Freezer
MDL Number: MFCD00057922
CAS Number: 50787-10-5

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR