Granite™ Black Nitrile Industiral Gloves
Supplier: AMERICAN NITRILE OPERATIONS, LLC
ANGR552 ANGR553 ANGR554 ANGR555 ANGR556
77888-578BX 12.99 USD
77888-578 77888-582 77888-584 77888-580 77888-576
Granite™ Black Nitrile Industiral Gloves
Gloves
Made in the USA, these premium nitrile gloves combine comfort, durability, and protection to optimize performance in hardworking applications.
  • Natural feel minimizes muscle fatigue
  • 6 mil fingertip thickness
  • Touchscreen friendly
  • ASTM F739 for chemical splash protection
  • ASTM F1342 for puncture resistance
The perfect balance of strength and flexibility, Granite™ gloves protect hands from harsh chemicals, abrasions, and more. Manufactured with a black finish to help identify grease, powder, and other substances.
Featuring compression fit technology and designed for manufacturing and industrial applications, these gloves are ideal for use in automotive, janitorial, police/law enforcement, warehouse, and construction industries.