Duplicate Sample Powder and Liquid Samplers, IV
Supplier: GLOBEPHARMA
M4.6.12.SS M4.2.1.SS M4.2.5.SS M4.3.1.SS M4.3.7.SS M4.4.1.SS M4.4.8.SS M4.5.1.SS M4.5.10.SS M4.6.1.SS
77887-060EA 4709.64 USD
77887-060 77888-052 77887-650 77887-026 77888-428 77888-416 77887-066 77887-884 77888-464 77888-118
Duplicate Sample Powder and Liquid Samplers, IV
Samplers Solid Samplers
Unit-dose and bulk powder, liquid, and semi-solid samplers available in various lengths and configurations. Duplicate sample devices are constructed so that each port holds two dies.
  • Precision sampling: Designed for accurate and repeatable sampling of powders from bulk containers, ensuring representative and consistent samples for quality control and testing
  • Pharmaceutical-grade construction: Made from high-quality 316L stainless steel or FDA-compliant materials, ensuring compatibility with GMP environments and resistance to corrosion or contamination
  • Ease of cleaning and sterilization: Engineered for quick disassembly and thorough cleaning, supporting clean-in-place (CIP) and autoclave sterilization procedures
  • Customizable and scalable: Can be tailored to specific sampling applications with adjustable volumes and configurations, ideal for R and D, pilot, and production-scale environments
  • Compliance ready: Supports regulatory compliance with FDA, USP, and ISO standards, making them suitable for validated sampling protocols in pharmaceutical and cleanroom operations
Global Pharma specializes in high-quality sampling solutions for the pharmaceutical, biotech, and related industries. The product portfolio includes unit dose and bulk powder, liquid, and semi-solid samplers, designed to meet stringent regulatory requirements. These samplers are available in a variety of configurations to ensure precise and contamination-free sample collection.
With a focus on innovation and compliance, Global Pharma’s samplers cater to diverse manufacturing environments, providing single-use and reusable options for enhanced process efficiency and sterility assurance. Their products are widely used in quality control, R and D, and production settings, making them a trusted choice for pharmaceutical manufacturers worldwide.