5-Formylfuran-2-boronic acid, pinacol ester 98%

Supplier: Ambeed
Warning

A693690-1G A693690-250MG A693690-25G A693690-5G
76831-574EA 12 USD
76831-574 77886-621 77886-622 77886-623
5-Formylfuran-2-boronic acid, pinacol ester 98%
5-Formylfuran-2-boronic acid, pinacol ester

Caution: for Research Use Only

Formula: C₁₁H₁₅BO₄
MDL Number: MFCD16295105
CAS Number: 273731-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR