(±)-2-Hydroxyisovaleric acid 97%

Supplier: Ambeed
Warning

A356201-1G A356201-100MG A356201-1MG A356201-25G A356201-5G A356201-10G A356201-100G
103826-162EA 40 USD
103826-162 103837-170 76840-308 76989-740 76989-742 77306-676 77306-678 77613-024 77885-939
(±)-2-Hydroxyisovaleric acid 97%
(±)-2-Hydroxyisovaleric acid

Caution: for Research Use Only

Formula: C₅H₁₀O₃
Boiling Pt: 237.6 °C (760 mm Hg)
Melting Pt: 86…87 °C
Flash Pt: 111.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00004242
CAS Number: 4026-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR