5-Amino-3-methylisoxazole 97%

Supplier: Ambeed
Warning

Synonyms: 3-Methylisoxazol-5-ylamine, 3-Methylisoxazol-5-amine

A274934-1G A274934-250MG A274934-25G A274934-5G A274934-10G
76875-170EA 6 USD
76875-170 76875-172 76875-174 76875-176 77883-757
5-Amino-3-methylisoxazole 97%
5-Amino-3-methylisoxazole

Caution: for Research Use Only

Formula: C₄H₆N₂O
MW: 98.1 g/mol
Boiling Pt: 194
Melting Pt: 84
MDL Number: MFCD00003151
CAS Number: 14678-02-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR