(6-Ethoxy-2,3-difluorophenyl)boronic acid 97%
Supplier: Ambeed
|
Warning
|
A258150-100MG
A258150-250MG
A258150-5G
103812-364EA
55
USD
103812-364
103852-238
76815-952
76815-954
77883-703
(6-Ethoxy-2,3-difluorophenyl)boronic acid 97%
(6-Ethoxy-2,3-difluorophenyl)boronic acid
Caution: for Research Use Only
Formula:
C₈H₉BF₂O₃ Boiling Pt: 332.3±52.0 °C (760 mm Hg) Flash Pt: 154.8 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD12026698 CAS Number: 1309980-95-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...